Filter thimbles with 38 mm flange for dietary fibre extraction, VitraPOR®
Lieferant: ROBU GLASFILTER
20682 20683 20684 20681 20680
ROBU20682EA 101 EUR
ROBU20682 ROBU20683 ROBU20684 ROBU20681 ROBU20680
Filter thimbles with 38 mm flange for dietary fibre extraction, VitraPOR®
Extraktoren Extraktionshülsen
Borosilicate glass 3.3.
  • Very good chemical resistance
  • Very good temperature resistance
Filter-Thimbles, CFE, with exact glass flange fits many current systems.