Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Rockland Immunochemicals
613-4528
ROCK613-4528EA 464 EUR
ROCK613-4528
Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
When choosing a secondary antibody, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:400 - 1:1600; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR