ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat]), Flüssigkeit ELISA Peroxidase-Substrat, anwendungsbereit

Lieferant: Rockland Immunochemicals

Synonyme: ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

ABTS-100
ROCKABTS-100EA 233 EUR
ROCKABTS-100
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat]), Flüssigkeit ELISA Peroxidase-Substrat, anwendungsbereit
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat])

ABTS ELISA Peroxidase Substrate (2,2'-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid)) is a chromogenic substrate use to visualize reactivity of certain enzymes, particularly in immunosorbent assays. ABTS ELISA Peroxidase Substrate is commonly used with peroxidases, but is amenable to many other enzymes. In the presence of active enzyme, the substrate becomes a soluble green product detectable at 405nm.


  • Ready-to-use substrate
  • Application: ELISA - 1X


This substrate finds use in many immunosorbent assays, particularly ELISA.


Storage Condition
Store container at 4° C prior to opening. Protect from moisture and light. No special shipping conditions or precautions are required.

Formel: C₁₈H₂₄N₆O₆S₄
Molecular Weight: 548.68 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00010404
CAS-Nummer: 30931-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR