Blocking reagents, Blotto
Lieferant: Rockland Immunochemicals
B501-0500
ROCKB501-0500EA
186
EUR
ROCKB501-0500
Blocking reagents, Blotto
Puffer
Elektrophorese-Puffer
Blockier-Puffer
Contains immunoanalytical grade non-fat dry milk. Blocks non specific binding of proteins in various immunological assays. Ideal for use in western blotting and ELISA methods.