BCIP/TNBT alkaline phosphatase substrate

Lieferant: Rockland Immunochemicals

Synonyme: TNBT (tetra nitro-blue tetrazolium chloride), BCIP (5-bromo-4-chloro-3-indolylphosphate toluidine salt)

BCBT-100
ROCKBCBT-100EA 233 EUR
ROCKBCBT-100
BCIP/TNBT alkaline phosphatase substrate
BCIP/TNBT

BCIP/TNBT Membrane Alkaline Phosphatase Substrate is an artificially manufactured chromogenic substrate for use in sensitive colorimetric assays. BCIP/TNBT can be utilized in ELISA, in situ hybridization, and IHC. Alkaline phosphatase hydrolyzes the BCIP to 5-bromo-4-chloro-3-indole, which is oxidized by oxygen in the atmosphere, yielding a blue dye. In the presence of TNBT, an insoluble dark blue precipitate is formed after a reduction reaction instead.


BCIP/TNBT Membrane Alkaline Phosphatase Substrate is a useful detection reagent for WB, IHC (immunohistochemistry), and in situ hybridisation.


Storage Condition: Store container at 4° C prior to opening. Protect BCIP/TNBT Membrane Alkaline Phosphatase Substrate from moisture and light.

Bestellen

Spezifikation Testergebnisse

pH 9.28 +/- 0.1
QC Raw Material PASS
Absorbance check of final product PASS
Performance Data per Immunoblot PASS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR