Fetal bovine serum, heat inactivated, certified, U.S. origin
Lieferant: Rockland Immunochemicals
FBS-01-0100
ROCKFBS-01-0100EA 341 EUR
ROCKFBS-01-0100
Fetal bovine serum, heat inactivated, certified, U.S. origin
Seren
Fatal bovine serum is used in a wide range of cell culture applications.
  • Sterile bacteriophage
  • Store at −20 °C
This bovine serum is certified to originate in the United States as heat inactivated, performance, mycoplasma, virus, bacteriophage and endotoxin tested.
Bitte beachten For in vitro diagnostic use only, not for human or animal therapeutic use.