Wasser, UltraPure, Filter sterile

Lieferant: Rockland Immunochemicals

Synonyme: Wasserstoffoxid, Hydrogenoxid

MB-009-1000 MB-009-4000
ROCKMB-009-1000EA 124 EUR
ROCKMB-009-1000 ROCKMB-009-4000
Wasser, UltraPure, Filter sterile
Wasser
Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR