TPE buffer, 10X tris-phosphate-EDTA, pH 8
Lieferant: Rockland Immunochemicals
MB-021
ROCKMB-021EA 195 EUR
ROCKMB-021
TPE buffer, 10X tris-phosphate-EDTA, pH 8
Puffer Elektrophorese-Puffer Laufpuffer
This product is a concentrated stock solution and should be diluted appropriately with distilled, deionised water or equivalent to its final working concentration. 10X Tris-Phosphate EDTA (TPE) consists of 0,9 M Tris-Phosphate, 0,02 M EDTA at a pH of 8,0. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionsed water.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Bitte beachten This product is for research use only and is not intended for therapeutic or diagnostic applications.