Deoxyribonuclease I
Lieferant: Rockland Immunochemicals
MB-101-0500
ROCKMB-101-0500EA
384
EUR
ROCKMB-101-0500
Deoxyribonuclease I
Enzyme
Deoxyribonuclease I (usually called DNase I), is an endonuclease coded by the human gene DNASE1. DNase I is a nuclease that cleaves DNA preferentially at phosphodiester linkages adjacent to a pyrimidine nucleotide, yielding 5'-phosphate-terminated polynucleotides with a free hydroxyl group on position 3', on average producing tetranucleotides. It acts on single-stranded DNA, double-stranded DNA, and chromatin.