Nuclease S1
Lieferant: Rockland Immunochemicals
MB-110-0010
ROCKMB-110-0010EA
201
EUR
ROCKMB-110-0010
Nuclease S1
Enzyme
Nuclease S1 is an endonuclease that is active against single-stranded DNA and RNA molecules. Although its primary substrate is single-stranded, it can also introduce single-stranded breaks in double-stranded DNA-RNA or DNA-RNA hybrids. Nuclease S1 is used in the laboratory as a reagent in nuclease protection assays. It is used in removing single stranded tails from DNA molecules to create blunt ended molecules and opening hairpin loops generated during synthesis of double stranded cDNA.