SAR Breathing airline filters, 6 outlet, Radex

Lieferant: RPB SAFETY
04-906
RPBS04-906EA 0 EUR
RPBS04-906
SAR Breathing airline filters, 6 outlet, Radex
Atemschutzmasken Atemschutz-Systeme
The RPB radex 6 outlet breathing air filter is designed to filter compressed breathing air of contaminants such as particulates down to 0,15 µm, odors, tastes, and moisture, in accordance with EN 12021 requirements.

  • Quick disconnect coupler that is compatibile with brands including Aro, CEJN, Dixion, and Rectus
  • 150 psi safety presure relief valve
  • Claw coupler port
  • Fitted with brass drain tap
  • Outlet pressure regulator provides the ability to adjust air pressure (0 to 125 psi range)
  • Strong, robust housing
  • Wall and floor mounting system
  • 6 outlet system to provide filtration to 6 seperate airlines
  • Operating temperature: −10 to 60 °C
  • Flow rate: 2123 L/min

The 6-stage stack filter cartridge offers a robust and cost-effective option for companies using compressed breathing air respirators, whether it be hard mounted in a paint room, or out on site, with the adaptable base mount.

Replacement filter cartridges have a replacement date sticker for maintenance scheduling. The filter does not remove gases such as carbon monoxide.

Standards: EN 14594:2005 3B/4B.

Lieferumfang: Package includes: 1×filter housing, 1×filter, 1×base/wall mount, 1×high flow pressure regulator and gauge, 6×couplers, 1×PRV and 1×drain tap.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR