The RPB Z-link respirator with FR face seal, is the world's most comfortable (fully padded) and versatile industry respirator. Providing the highest respiratory protection available (TH3), the Z-Link system also provides hard hat protection, eye and face protection and hearing protection.
- Visor hinges open for easy communication with team (outside of respiratory hazards)
- Air flow and distribution is fully adjustable with the air deflector system to suit personal preferences
- Fixed jaw ensures secure cape attachment
- Vision-Link™ light mount point
- ANSI Z89.1 certified head protection (EN 397)
- Padded interior with adjustable ratchet system ensures a perfect customized fit that supports movement, increasing comfort and productivity
- Three cape materials available: Tychem® 2000, Tychem® 4000, or Zytec® FR
- Operating temperature: −10 to +60 °C
- Noise rating: NRR 25 (side pads), NRR 41 (ear defenders and earplugs)
Optimum vision and high velocity impact protection with large optically-clear safety lens (replaceable spare part). Choice of 5 antifog and scratch resistant ANSI Z87+ (EN 166) lenses: premium clear, green tint (shade 5), yellow tint, black tint and mirror tint.
The Z-link is designed to be able to perform all tasks within a business, including general RPE, welding, grinding, painting/coating and now high heat foundry works. This allows companies to standardize systems and reduce alternate system and brands spare part stocking and usage.
To achieve this, the Z-link offers a wide range of market leading out of the box accessory items to ensure user comfort, safety and productivity. These include:
• Clip on weld visor with 1:1:1 ADF (16-670)
• Chin mounted helmet light (16-901)
• In-helmet communication system (16-922)
• Quiet-Link ear defenders (18-533)
• Comfort fit side pads (as standard)
• IR5 visors (16-810-IR5, 16-675-IR5)
• High heat foundry kits, including gold visors (16-674, 16-744, 04-955, 16-675-GT)
Standards: EN 12941:1998+A2:2008 TH3/2 P R SL, EN 397:2012+A1:2012, EN 166:2001 Class 1BT 9 3.
Lieferumfang: Includes PX5 PAPR breathing tube. PAPR sold separately.