Stainless steel, Drahtgeflecht für Gasbrenner
Lieferant: RSG
247.875
247.855
247.835
247.815
247.895
RSGA247.875EA
52.6
EUR
RSGA247.875
RSGA247.855
RSGA247.835
RSGA247.815
RSGA247.895
Stainless steel, Drahtgeflecht für Gasbrenner
Stainless steel
Wire mesh out of stainless steel wire with folded-up borders
- Stainless steel 1.4301
- with folded up edge
- mesh: 0.6 x 2.0 mm