Stainless steel, Drahtgeflecht für Gasbrenner
Lieferant: RSG
247.875 247.855 247.835 247.815 247.895
RSGA247.875EA 52.6 EUR
RSGA247.875 RSGA247.855 RSGA247.835 RSGA247.815 RSGA247.895
Stainless steel, Drahtgeflecht für Gasbrenner
Stainless steel
Wire mesh out of stainless steel wire with folded-up borders
  • Stainless steel 1.4301
  • with folded up edge
  • mesh: 0.6 x 2.0 mm