Beaker tongs, stainless steel
Lieferant: RSG
80.043.330
RSGA80.043.330EA 42.1 EUR
RSGA80.043.330
Beaker tongs, stainless steel
Zangen
Beaker/cup tongs are made from stainless steel 1.4301.
  • Electrolytically polished
  • Jaws wrapped with organic fleece