Sulphobromophthalein sodium salt hydrate, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

S0252-5G
S0252-5GEA 546 EUR
S0252-5G
Sulphobromophthalein sodium salt hydrate, Sigma-Aldrich®
Sulphobromophthalein sodium salt hydrate

Sulfobromophthalein disodium salt hydrate has been used as a positive control to study the interaction of 11-keto-β-boswellic acid (KBA) and 3-acetyl-11-keto-β-boswellic acid (AKBA) with organic anion transporter OATP1B3.

Formel: C₂₀H₈Br₄O₁₀S₂Na₂·xH₂O
Molecular Weight: 837.98 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150017
CAS-Nummer: 71-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38571c3ab5c9565f238c47792d4bbc6b3c1063bc&vid=55951e84fda4fa2ad68ba2919cd7d5594ac8b894&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR