Rat serum
Lieferant: Biowest
S2150-100 S2150-500 S2150-050 S2150-010 S2150-020
S2150-100EA 0 EUR
S2150-100 S2150-500 S2150-050 S2150-010 S2150-020
Rat serum
Seren
Rat serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Rat serum is used as a blocking agent in immunoassays and controls.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable