Dog plasma
Lieferant: Biowest
S2930-100
S2930-100EA 0 EUR
S2930-100
Dog plasma
Plasma
Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.