Sudan IV, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Sudan® IV (Scharlachrot), 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol, Oil Red BB, Sudan M Rot 380, Oil Red IV, Solvent Red 24, 1-([2-Methyl-4-[(2-methylphenyl)diazenyl]phenyl]diazenyl)naphthalen-2-ol, Oil Red, Fat Red B, Scarlet Red, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach

M-Clarity™ Qualitätsstufe = MQ200

S4261-25G
S4261-25GEA 143 EUR
S4261-25G
Sudan IV, Sigma-Aldrich®
Sudan IV
Formel: C₂₄H₂₀N₄O
Molecular Weight: 380.45 g/mol
Schmelzpunkt: 188
Lagertemperatur: Raumtemperatur
MDL: MFCD00003893
CAS-Nummer: 85-83-6
EINECS: 201-635-8
Merck Index: 12,08538
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ab94a35cf9f6677fbfc347163652a8388ebf0d4&vid=76c57581d44fc7471d7e20a39e29260783782dd9&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR