Sodium diatrizoate hydrate ≥98%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Amidotrizoate sodium salt hydrate, Sodium 3,5-diacetamido-2,4,6-triiodobenzoate hydrate, 3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid sodium salt hydrate, 3,5-Diacetamido-2,4,6-triiodobenzoic acid sodium salt hydrate, Diatrizoate sodium hydrate, Diatrizoic acid sodium salt hydrate

M-Clarity™ Qualitätsstufe = MQ200

S4506-100G S4506-10G S4506-250G S4506-500G S4506-50G
S4506-100GEA 343 EUR
S4506-100G S4506-10G S4506-250G S4506-500G S4506-50G
Sodium diatrizoate hydrate ≥98%, Sigma-Aldrich®
Sodium diatrizoate hydrate

Sodium diatrizoate hydrate has been used in spore purification. It has also been used in the isolation of plasma lymphocytes, monocytes, and dendritic cells. Sodium diatrizoate is also suitable as a density gradient reagent in blood cell separation. It has also been utilised as a radiological contrast agent.

Formel: C₁₁H₈I₃N₂NaO₄·xH₂O
Molecular Weight: 635.898 g/mol (anhy)
Schmelzpunkt: 261 °C
Lagertemperatur: Room Temperature
CAS-Nummer: 737-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a548cf4b37fe1c8932dce4cdca342f7e87d65b84&vid=692f1589e4960c98bc3265f05db29c0c50cb2979&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Off White
Appearance (Form) Powder
1H NMR Spectrum Conforms to Structure
Water (by Karl Fischer) ≤11.0 %
Titration
Anhydrous
≥98.0 %
Solubility (Colour) Colorless to Faint Yellow
Solubility (Turbidity)
350 mg/ml, H2O
Clear to Slightly Hazy


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR