Sulphochlorophenol S sodium calcium salt, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

S5752-500MG
S5752-500MGEA 72.6 EUR
S5752-500MG
Sulphochlorophenol S sodium calcium salt, Sigma-Aldrich®
Sulphochlorophenol S sodium calcium salt

Sulfochlorophenol S sodium calcium salt, (2,7-Bis(2-hydroxy-3-sulfo-5-chlorophenylazo)-1,8-di-hydroxynaphthalene-3,6-disulfonic acid)

Formel: C₂₂H₁₄Cl₂N₄O₁₆S₄
Lagertemperatur: Kühlschrank
MDL: MFCD00151404
CAS-Nummer: 108321-09-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=306601c7e932affce8433c20435d56c9036bdd17&vid=145389471f162592df74ed44b00386f5307821d9&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR