Natriumhydrogencarbonat 99,7 ACS, Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
Natriumbicarbonat
S6014-12KG
S6014-1KG
S6014-2.5KG
S6014-500G
S6014-5KG
S6014-6X500G
S6014-12KGEA
617.07
EUR
S6014-12KG
S6014-1KG
S6014-2.5KG
S6014-500G
S6014-5KG
S6014-6X500G
Natriumhydrogencarbonat 99,7 ACS, Fluka™
Natriumhydrogencarbonat
Formel:
NaHCO₃ Molecular Weight: 84.01 g/mol Schmelzpunkt: 270 Zersetzung Dichte: 2.2 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003528 CAS-Nummer: 144-55-8 EINECS: 205-633-8 Merck Index: 13,08655 |
|
Spezifikation Testergebnisse
Identity (IR) | complying |
Assay (excluding dried substance) | 99.7 - 100.3 % |
Water insoluble matter | Max. 0.015 % |
Calcium (Ca) | Max. 0.02 % |
Iron (Fe) | Max. 0.001 % |
Potassium (K) | Max. 0.005 % |
Magnesium (Mg) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 0.003 % |
Phosphate (PO₄) | Max. 0.001 % |
S-compounds (as SO₄) | Max. 0.003 % |
Ammonium (NH₄) | Max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...