SynaptoGreen™ C4 ≥95% (durch HPLC), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Pyridinium, 4-[2-[4-(dibutylamino)phenyl]ethenyl]- 1-[3-(triethylammonio)propyl]- , dibromide, SynaptoGreen™ C4, FM1-43
S6814-5MG
S6814-5MGEA
622
EUR
S6814-5MG
SynaptoGreen™ C4 ≥95% (durch HPLC), Sigma-Aldrich®
N-(3-Triethylammoniumpropyl)-4-(4-(dibutylamino)styryl)pyridinium dibromide (MM 1-43)
SynaptoGreen(TM) C4. ≥95% (HPLC), solid (FM1-43; N-(3-triethylammoniumpropyl) -4-(4-(dibutylamino)styryl) pyridinium dibromide; Pyridinium, 4-[2-[4-(dibutylamino)phenyl]ethenyl]- 1-[3-(triethylammonio)propyl]- , dibromide)
Formel:
C₃₀H₄₉Br₂N₃ Molecular Weight: 611,54 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00467815 CAS-Nummer: 149838-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...