Stopcocks, three-way, Supelco®
Lieferant: Merck
S7521-10EA
S7521-10EAEA 122 EUR
S7521-10EA
Stopcocks, three-way, Supelco®
Hähne
Made of nylon.
  • Good chemical resistance
  • Good temperature resistance
Features three-way control with female luer-lock inlet and male luer-lock outlet ports.