Sulphorhodamine 101 ~95 % (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ100

S7635-100MG S7635-500MG S7635-50MG
S7635-100MGEA 287 EUR
S7635-100MG S7635-500MG S7635-50MG
Sulphorhodamine 101 ~95 % (Farbstoffgehalt), Sigma-Aldrich®
Sulphorhodamine 101

Sulforhodamine 101 is a red fluorescent dye. It is water-soluble, amphoteric rhodamine.


  • Sulforhodamine 101 has been used
  • to label astrocytes
  • in quantitative polymerase chain reaction (PCR)
  • to determine the degree of disintegration in tissue sample


Sulforhodamine 101 can be specifically used as a marker of astroglia in the neocortex. It is commonly used for brain imaging. Studies suggest that sulforhodamine 101 may be used as an epileptogenic agent.

Formel: C₃₁H₃₀N₂O₇S₂
Lagertemperatur: Room Temperature
MDL: MFCD00012407
CAS-Nummer: 60311-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8806caeca19e94722ec826972cbad139f767211f&vid=13577b91b027b18a7d575b02df247dd61c11ac0e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Dark Green to Brown to Black
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Carbon (dry basis) 57.5 - 65.0 %
Nitrogen (dry basis) 4.2 - 5.0 %
Wavelength 573 - 579 nm
Extinction Coefficient ≥105000
In Ethanol
Water (by Karl Fischer) %
Solubility (Colour) Conforms
Dark Pink (C = 1mg/ml; Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR