Natriumhydroxid, anhydrous 98 Acidimetry, pellets, BioXtra

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Caustic soda

S8045-1KG
S8045-1KGEA 143.56 EUR
S8045-1KG
Natriumhydroxid, anhydrous 98 Acidimetry, pellets, BioXtra
Sodium hydroxide

  • Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
  • Includes application-specific formulations, such as tablets and prepared solutions.

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1.390 °C (1.013 hPa)
Schmelzpunkt: 319 °C
Dichte: 2.13 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
REACH: 01-2119457892-27
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

Insoluble matter Max. 0.1 %
Solution 1 M in H₂O complying
Assay (acidimetric) Min. 98.0 %
Aluminium (Al) Max. 0.0005 %
Calcium (Ca) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.02 %
Magnesium (Mg) Max. 0.0005 %
Phosphorus (P) Max. 0.0005 %
Lead (Pb) Max. 0.001 %
Zinc (Zn) Max. 0.0005 %
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.05 %
Form pellets (anhydrous)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR