Saffran, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ200

S8381-1G S8381-5G
S8381-1GEA 129 EUR
S8381-1G S8381-5G
Saffran, Sigma-Aldrich®
Saffran
CAS-Nummer: 84604-17-1
EINECS: 283-295-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR