Sampling tips for UD sleeve sampler
Lieferant: Sampling Systems
1500W-14 1500W-13 1500W-15
SAMP1500W-14EA 204 EUR
SAMP1500W-14 SAMP1500W-13 SAMP1500W-15
Sampling tips for UD sleeve sampler
Probenehmer Probenehmer für Feststoffe
The UD Sleeve sampler is perfect for small volume powder sampling.
  • Extensively used in the pharmaceutical industry
  • Accurate sampling of free-flowing powders
  • Extension rods are avialble to increase the length of the sampler
The UD Sleeve Sampler is ideal for accurate sampling of small volumes of free-flowing powders. It is made from 316L stainless steel and available with a range of tips, enabling samples sizes between 0,1 and 2,5 ml. Various accessories are available, such as extension rods which anable the operator to extend the length of the sampler.