Powder lances
Lieferant: Sampling Systems
2250A-1000 2250A-2000 2250A-600
SAMP2250A-1000EA 677 EUR
SAMP2250A-1000 SAMP2250A-2000 SAMP2250A-600
Powder lances
Probenehmer Probenehmer für Feststoffe
The powder lance can be used to manually take large volume samples.
  • Cross sectional sampling without damage
  • Mostly used with cohesive powders
  • Horizontal use through a sack or elsewhere vertically
Suitable for use in the pharmaceutical industries.