Heavy-duty hand reel
Lieferant: Sampling Systems
5070W-01
SAMP5070W-01EA 136 EUR
SAMP5070W-01
Heavy-duty hand reel
Probenehmer Probenehmer für Flüssigkeiten
Essential for sampling with Sub Samplers and bailers.
  • High capacity reel
  • Built-in lcoking mechanism and hanging hooks
A popular range of hand reels and cables are essential when sampling with Sub Samplers and bailers.