SteriWare Cake Cutters
Lieferant: Sampling Systems
8765H-4050 8765H-4050S
SAMP8765H-4050EA 158 EUR
SAMP8765H-4050 SAMP8765H-4050S
SteriWare Cake Cutters
Probenehmer Probenehmer für Feststoffe
These circular cutters are perfect for taking samples of cake like material.
  • Made from FDA and EC compliant HDPE
  • 100% cleanroom manufactured and packed
  • Choice of sterile and non sterile
These individually bagged samplers are perfect for taking small 2 to 3 cm samples of cake-like material typically from in trays.