Ladles, USP Class VI, double-bagged
Lieferant: Sampling Systems
9053H-50S
SAMP9053H-50SEA 235 EUR
SAMP9053H-50S
Ladles, USP Class VI, double-bagged
Schaufeln
These sterile HDPE ladles are great for taking fast liquid samples. The entire range have a slim line design so they can work through 2" bungs. They are commonly used for taking liquid samples from tubs, containers, bins etc.
  • Double-bagged
  • Produced in Class 100,000 cleanroom, packed in Class 10,000 cleanroom
The ladles are supplied double bagged as part of our SteriWare Ultra range. The box is also lined with a further bag for extra protection.
Standards: USP Class VI Medical grade polymer is FDA and EC compliant, tested to USP661. Certified BSE/TSE free.
Verpackung: Supplied in a lined box.