Ladles
Lieferant: Sampling Systems
A324-50 A324-120 A324-150 A324-200 A326-150
SAMPA324-50EA 35.5 EUR
SAMPA324-50 SAMPA324-120 SAMPA324-150 SAMPA324-200 SAMPA326-150
Ladles
Schaufeln
These ladles made from hygienic stainless steel are ideal for sampling liquid products. The ladles have a crevice free construction so that they can be easily cleaned between batches.
  • Crevice free construction allows easy cleaning between batches
  • Satin finish