X-ray and metal detectable scoops, SteriWare®
Lieferant: Sampling Systems
M1000B-1000 M1000B-125 M1000B-250 M1000B-2500 M1000B-30 M1000B-500 M1000B-60
SAMPM1000B-1000EA 95.9 EUR
SAMPM1000B-1000 SAMPM1000B-125 SAMPM1000B-250 SAMPM1000B-2500 SAMPM1000B-30 SAMPM1000B-500 SAMPM1000B-60
X-ray and metal detectable scoops, SteriWare®
Schaufeln
The X-ray and metal detectable scoops are ideal for use in food and pharmaceutical production. They are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the powder in line with HACCP guidelines.
  • Robust PP construction, provides excellent chemical resistance to many products
  • Available in volumes from 30 to 2500 ml
  • Full batch traceability
To ensure the cleanliness of the scoops they are manufactured and packed in a Cleanroom. They can be used as a single use disposable product straight from the bag or reused. The robust nature of these scoops means that they give many years of service.
Standards: These PP scoops conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Verpackung: Supplied individually bagged.