X-ray and metal detectable liquid funnels
Lieferant: Sampling Systems
M1023B-100
SAMPM1023B-100EA 22 EUR
SAMPM1023B-100
X-ray and metal detectable liquid funnels
Trichter Filtertrichter Filtertrichter
These X-ray and metal detectable liquid funnels are perfect for use in the laboratory and production. These funnels are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the material in line with HACCP guidelines.
  • Robust PP construction, provides excellent chemical resistance to many products
  • Full batch traceability
  • Easy to use
To ensure the cleanliness of the funnels, they are manufactured and packed in a cleanroom. They can be used as a single use disposable product straight from the bag or reused.
Standards: These PP funnels conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Verpackung: Supplied individually bagged.