Belüftungsfilter, Midisart® 2000
Lieferant: Sartorius
17805-E
SARP17805-EEA 129 EUR
SARP17805-E
Belüftungsfilter, Midisart® 2000
Filter Belüftungsfilter
Venting filters for the sterile venting of small fermenters and culture vessels.
Hydrophobic PP reinforced PTFE membrane, PP housing