Cassettes, Sartocon®
Lieferant: Sartorius
3021443906E-SG
SARP3021443906E-SGEA 4760 EUR
SARP3021443906E-SG
Cassettes, Sartocon®
Filter Tangentialfluß-Filtrationskassetten
Hydrosart® membranes are stabilised cellulose based membranes that have been optimised for the biotechnological and pharmaceutical industry. It is a stable polymer that features a broad pH range. Hydrosart® is also extremely hydrophilic, making it non-protein binding, virtually non-foul and has extremely high flux. Membrane regeneration, storage and depyrogenation can be accomplished by using NaOH even at elevated temperatures. These features make Hydrosart® an ideal membrane for biological applications.
PESU (polyethersulfone) membranes are membrane polymers that are well established in the biotechnological and pharmaceutical industries. It is a stable polymer that features a broad pH and temperature range. Its wide temperature range makes it possible to sterilise the membrane by either steam or autoclaving. Membrane regeneration, storage and depyrogenation can be accomplished by using NaOH even at elevated temperatures. Because of these features, the PESU membrane is ideally suited for biotechnological applications.