For the reduction of viruses in biopharmaceutical feed streams. The Virosart® HF Module with 200cm² filtration area for scale up runs as well as smaller production volumes. Adapter type: inlet ¾” Sanitary, outlet:Hose barb, sterile delivery 1 * 1 ST
Lieferant: Sartorius
For the reduction of viruses in biopharmaceutical feed streams. The Virosart® HF Module with 200cm² filtration area for scale up runs as well as smaller production volumes. Adapter type: inlet ¾” Sanitary, outlet:Hose barb, sterile delivery 1 * 1 ST