Filtration capsules, Sartolon® MidiCaps
Lieferant: Sartorius
5105307H9-OO-A
SARP5105307H9-OO-AEA 812 EUR
SARP5105307H9-OO-A
Filtration capsules, Sartolon® MidiCaps
Filter Membranfilterkapseln
Ready-to-use disposable capsules for sterile filtration in small and pilot scale production. Designed for broadest chemical compatibility for specific applications in the pharmaceutical and chemical industry. Suited for sterilising grade filtration of a broad range of solvents and liquids containing solvents.
  • Broad chemical compatibility
  • Flexible, scalable and cost saving
Nylon membrane; support fleece, core and end caps: PP; O-rings: silicone; housing: PP
Sartolon® MidiCaps are designed, developed and manufactured in accordance with a ISO 9001 certified Quality Management System. A Validation Guide is available for compliance with regulatory requirements.