Filtration capsules, Sartopore® 2 XLG
Lieferant: Sartorius
5445307G7-SO-A 5445307G0-SO-V 5445307G8-SO-A 5441307G4-SS-B
SARP5445307G7-SO-AEA 671 EUR
SARP5445307G7-SO-A SARP5445307G0-SO-V SARP5445307G8-SO-A SARP5441307G4-SS-B
Filtration capsules, Sartopore® 2 XLG
Filter Membranfilterkapseln
Sartopore® 2 XLG capsules are self contained filter units that are especially designed for sterilising grade filtration in special applications of cell culture processes. Their unique heterogeneous double layer PES membrane combination is specifically developed to deal with the broad variety of contaminants in up- and downstream processing of biotechnology applications. They provide consistently high total throughput performance for biological fluid streams independent from media and process variations.
  • Easy to use
  • Flexible and cost saving
  • Ideal for scale-up and scale-down
  • Compatible
More Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable
Max. Druck 5 bar @ 20 °C; 2 bar @ 80 °C 4 bar @ 20 °C; 2 bar @ 80 °C