Sample vials for coulter counter
Lieferant: SARSTEDT
73.650.500
SARS73.650.500EA 50.3 EUR
SARS73.650.500
Sample vials for coulter counter
Vials
Sample vials for Coulter Counter with enclosed caps. Vials are made from PS, caps from LDPE.
  • Flat base
Verpackung: 250 vials and caps are packed in the bag.