Transport container, round
Lieferant: SARSTEDT
78.898 78.890 78.892 78.895 78.896 78.897 78.898.300
SARS78.898EA 101 EUR
SARS78.898 SARS78.890 SARS78.892 SARS78.895 SARS78.896 SARS78.897 SARS78.898.300
Transport container, round
Probenbehälter Universalbehälter
These mailing containers are made of PP. Suitable for the transport of diagnostic samples.
  • Leakproof at a pressure difference of 95 kP
  • Without cap
Standards: Compliance with the P650/ADR packaging instructions.