Cell culture dishes for adherent cells, Sarstedt SUREGrip design
Lieferant: SARSTEDT
83.3902
SARS83.3902EA 561 EUR
SARS83.3902
Cell culture dishes for adherent cells, Sarstedt SUREGrip design
Zellkulturschalen Zell- und Gewebekultur-Platten
PS. Standard TC treated dishes with convenient handling due to a raised ring around the circumference of the dish base, enabling an easy and secure grip of both the dish and the lid, even from a whole stack.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable