Serological pipettes
Lieferant: SARSTEDT
86.1252.025 86.1254.025 86.1254.001 86.1251.001 86.1253.001 86.1685.001 86.1251.025 86.1252.001 86.1253.025 86.1685.020
SARS86.1252.025EA 0 EUR
SARS86.1252.025 SARS86.1254.025 SARS86.1254.001 SARS86.1251.001 SARS86.1253.001 SARS86.1685.001 SARS86.1251.025 SARS86.1252.001 SARS86.1253.025 SARS86.1685.020
Serological pipettes
Pipetten Serologische Pipetten
PS, graduated, sterile.
  • Plugged
  • Easy identification by colour code
  • Larger pipetting volume due to negative graduation
  • Optimised mouth piece for universal fit in most common pipettors
  • Easy to open, static-free packaging