Vacuum hand pump
Lieferant: Sartorius
16673
SART16673EA 239 EUR
SART16673
Vacuum hand pump
Pumpen Vakuum-Handpumpen
The hand operated vacuum pump made of PVC is used as practical vacuum source for outside of a laboratory.
Lieferumfang: Supplied completely with gauge, vacuum release lever and a clear plastic tubing (600 mm).