Filter capsules, Sartofluor® LG MaxiCaps®
Lieferant: Sartorius
5187307T1-SS
SART5187307T1-SSEA 569 EUR
SART5187307T1-SS
Filter capsules, Sartofluor® LG MaxiCaps®
Filter Membranfilterkapseln
Sartofluor® LG MaxiCaps® are specially developed for liquid filtration. The modified construction is beneficial with regard to hydrodynamic flow and it is especially recommended for solvent filtration but can also be used as an air and gas filter. It improves the process security of sterile filtration of aggressive media (acids and bases) and solvents.
  • Single layer construction provides maximum flow rate
PTFE membrane; support fleece, core and end caps: PP; O-rings: EPDM; housing: PP
Standards: Sartofluor® LG MaxiCaps® are designed, developed and manufactured in accordance with a ISO 9001 certified Quality Management System. A Validation Guide is available for compliance with regulatory requirements.