Glass Fibre Extraction Thimbles, G400
Lieferant: Sartorius
FT-1204-030100 FT-1204-043123 FT-1204-022080 FT-1204-025100 FT-1204-019090 FT-1204-026060 FT-1204-033094
SARTFT-1204-030100EA 220 EUR
SARTFT-1204-030100 SARTFT-1204-043123 SARTFT-1204-022080 SARTFT-1204-025100 SARTFT-1204-019090 SARTFT-1204-026060 SARTFT-1204-033094
Glass Fibre Extraction Thimbles, G400
Extraktoren Extraktionshülsen
These glass fibre thimbles are primarily designed to fit the Soxhlet apparatus and potential applications include environmental monitoring (e.g. separation of dust, aerosols, gas or air streams) and food control (e.g. extraction of fats, emulsifiers or additives).
  • Composed of glass fibre cellulose for uncontaminated results
  • Engineered for superior mechanical stability to withstand rigorous use
  • Glass fibre thimbles offer remarkable resistance to high temperatures
  • Designed for a precise fit across a diverse range of extractors
  • Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
They are well suited to high temperature applications and are free of binders with high purity.