Filter papers, non woven
Lieferant: Sartorius
FT-3-01304-025
SARTFT-3-01304-025EA 176 EUR
SARTFT-3-01304-025
Filter papers, non woven
Filter Filterpapiere Qualitatives Filterpapier
These non woven grades are made of rayon. Ideal for the filtration or prefiltration of viscous solutions containing particles visible with the naked eye.