Real-Time PCR kit for detection of mycoplasma, Microsart® ATMP Mycoplasma
Lieferant: Sartorius
SMB95-1003
SARTSMB95-1003EA 554 EUR
SARTSMB95-1003
Real-Time PCR kit for detection of mycoplasma, Microsart® ATMP Mycoplasma
Mikrobiologische Testsysteme PCR-Tests
The Microsart® ATMP Mycoplasma detection kit is especially designed for an easy and fast detection of mycoplasma in cell-based therapeutics like autologous chondrocyte transplants (ATMPs –advanced therapy medical products). Due to the short shelf life of these therapeutics, a fast result is needed. The PCR-technology makes it possible to reduce the time-to-result to only 3 hours.
The Microsart® ATMP Mycoplasma can be easily implemented because no specific hardware is needed. The kit can be used with any type of real-time PCR cycler that is able to detect the fluorescence dyes FAM™ and ROX™. The total testing procedure takes only 3 hours.
The kit contains all essential components in a function tested ready-to-use master mix, including the polymerase.
Standards: Validated according to EP 2.6.7.