Jars, wide mouth, PFA
Lieferant: SAVILLEX
100-0060-01 100-0360-01 100-2000-01
SAVI100-0060-01EA 44.8 EUR
SAVI100-0060-01 SAVI100-0360-01 SAVI100-2000-01
Jars, wide mouth, PFA
Dosen Dosen, Universal
Jars, made of molded translucent PFA.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Screw caps available seperately
For sample collection, storage and transport. With radiused inside design for easy sample removal.