11ml Aufschnappöffnung Service temperature range of PFA is -328F to 500F (-200C to 260C) Will fit most standard microcentrifuge models RCF up to 10,000 g 1 * 1 ST
Lieferant: SAVILLEX
11ml Aufschnappöffnung Service temperature range of PFA is -328F to 500F (-200C to 260C) Will fit most standard microcentrifuge models RCF up to 10,000 g 1 * 1 ST